3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
Thermo Scientific Chemicals

3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%

CAS: 2478-38-8 | C10H12O4 | 196.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 115540050
Price (USD)
165.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
165.00
Each
Chemical Identifiers
CAS2478-38-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point123°C to 128°C
Titration with NaOH>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only