2,2-二甲氧基丙烷
2,2-二甲氧基丙烷
2,2-二甲氧基丙烷
Thermo Scientific Chemicals

2,2-二甲氧基丙烷

CAS: 77-76-9 | C5H12O2 | 104.15 g/mol
Have Questions?
更改视图buttonViewtableView
数量:
500 mL
1 L
2.5 L
10 L
包装:
查看全部
Metal Drum
货号 115630100
价格(TWD)
-
数量:
10 L
包装:
Metal Drum
请求批量或定制报价
化学标识符
CAS77-76-9
IUPAC Name2,2-dimethoxypropane
Molecular FormulaC5H12O2
InChI KeyHEWZVZIVELJPQZ-UHFFFAOYSA-N
SMILESCOC(C)(C)OC
查看更多内容
规格Specification Sheet规格表
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
查看更多内容
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only