Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Thermo Scientific Chemicals

Dimethyl acetylenedicarboxylate, 98%

CAS: 762-42-5 | C6H6O4 | 142.11 g/mol
数量:
5 g
25 g
100 g
500 g
製品番号(カタログ番号) 115711000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS762-42-5
IUPAC Name1,4-dimethyl but-2-ynedioate
Molecular FormulaC6H6O4
InChI KeyVHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC(=O)OC
さらに表示
Appearance (Form)Liquid
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only