Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Thermo Scientific Chemicals

Dimethyl acetylenedicarboxylate, 98%

CAS: 762-42-5 | C6H6O4 | 142.11 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
產品號碼 115715000
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS762-42-5
IUPAC Name1,4-dimethyl but-2-ynedioate
Molecular FormulaC6H6O4
InChI KeyVHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC(=O)OC
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only