4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
Thermo Scientific Chemicals

4-Dimethylaminoantipyrine, 97%

CAS: 58-15-1 | C13H17N3O | 231.30 g/mol
数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 115741000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS58-15-1
IUPAC Name4-(dimethylamino)-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular FormulaC13H17N3O
InChI KeyRMMXTBMQSGEXHJ-UHFFFAOYSA-N
SMILESCN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
さらに表示
Melting point105°C to 110°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only