4-Dimethylaminobenzoic acid, 98%
4-Dimethylaminobenzoic acid, 98%
4-Dimethylaminobenzoic acid, 98%
Thermo Scientific Chemicals

4-Dimethylaminobenzoic acid, 98%

CAS: 619-84-1 | C9H11NO2 | 165.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
產品號碼 115761000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS619-84-1
IUPAC Name4-(dimethylamino)benzoic acid
Molecular FormulaC9H11NO2
InChI KeyYDIYEOMDOWUDTJ-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(O)=O
檢視更多
規格Specification Sheet規格表
Melting point238°C to 242°C
Water=<0.5 % (K.F.)
Titration with NaOH>=97.5 % (On dry substance)
Appearance (Color)White to beige to grey-green
Appearance (Form)Crystalline powder
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only