2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals

数量:
5 g
25 g
100 g
製品番号(カタログ番号) 115810250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13242-44-9
IUPAC Namehydrogen 2-(dimethylamino)ethane-1-thiol chloride
Molecular FormulaC4H12ClNS
InChI KeyNRVFDGZJTPCULU-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCS
さらに表示
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration Iodimetric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only