Dansyl chloride, 98%
Dansyl chloride, 98%
Dansyl chloride, 98%
Thermo Scientific Chemicals

Dansyl chloride, 98%

CAS: 605-65-2 | C12H12ClNO2S | 269.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 115850250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS605-65-2
IUPAC Name5-(dimethylamino)naphthalene-1-sulfonyl chloride
Molecular FormulaC12H12ClNO2S
InChI KeyXPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Melting point70°C to 74°C
Appearance (Color)Yellow to light orange
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only