3-Dimethylaminopropiophenone hydrochloride, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropiophenone hydrochloride, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropiophenone hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Dimethylaminopropiophenone hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
11589025025 gGlass bottle
Catalog number 115890250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS879-72-1
IUPAC Namehydrogen 3-(dimethylamino)-1-phenylpropan-1-one chloride
Molecular FormulaC11H16ClNO
InChI KeyDKNDBIIKSJWQFL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point152°C to 155°C
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only