2,6-Dimethylaniline, 99%
2,6-Dimethylaniline, 99%
2,6-Dimethylaniline, 99%
Thermo Scientific Chemicals

2,6-Dimethylaniline, 99%

CAS: 87-62-7 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 115940050
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.90
Save 4.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.90
Save 4.25
Each
Chemical Identifiers
CAS87-62-7
IUPAC Name2,6-dimethylaniline
Molecular FormulaC8H11N
InChI KeyUFFBMTHBGFGIHF-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.5591 to 1.5611 (20°C, 589 nm)
Appearance (Color)Clear yellow to red-brown
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only