2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
Thermo Scientific Chemicals

2,3-Dimethylbutane, 98+%

CAS: 79-29-8 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116100010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS79-29-8
IUPAC Name2,3-dimethylbutane
Molecular FormulaC6H14
InChI KeyZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILESCC(C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3740 to 1.3760 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only