Dimethyl carbonate, 99%
Dimethyl carbonate, 99%
Dimethyl carbonate, 99%
Dimethyl carbonate, 99%
Thermo Scientific Chemicals

Dimethyl carbonate, 99%

CAS: 616-38-6 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 116120025
Price (USD)
202.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
202.00
Each
Chemical Identifiers
CAS616-38-6
IUPAC Namedimethyl carbonate
Molecular FormulaC3H6O3
InChI KeyIEJIGPNLZYLLBP-UHFFFAOYSA-N
SMILESCOC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<30 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.1 % (K.F.)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
RUO – Research Use Only