Taq DNA Polymerase (Brasil), 5,000 units - FAQs
View additional product information for Taq DNA Polymerase (Brasil) - FAQs (11615044)
1 product FAQs found
Which PCR polymerase do you offer that has the highest fidelity?
We offer Platinum SuperFi II DNA Polymerase that displays more than 300 times higher fidelity than Taq Polymerase.