N,N'-Dimethylethylenediamine, tech., 85%
N,N'-Dimethylethylenediamine, tech., 85%
N,N'-Dimethylethylenediamine, tech., 85%
Thermo Scientific Chemicals

N,N'-Dimethylethylenediamine, tech., 85%

CAS: 110-70-3 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 116190250
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Chemical Identifiers
CAS110-70-3
IUPAC Namemethyl[2-(methylamino)ethyl]amine
Molecular FormulaC4H12N2
InChI KeyKVKFRMCSXWQSNT-UHFFFAOYSA-N
SMILESCNCCNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=80 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only