Interested in a bulk or custom version of this product? Request pricing.

UCHL1 Recombinant Human Protein, N-His Tag

Catalog number: 11663H07E5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L1 (UCHL1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the native human UCHL1 (NP_004172.2) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus.

Activity: Measured by its ability to hydrolyze Ubiquitin AMC. The specific activity is >140 pmoles/min/µg.

Formulation: 20 mM Tris, 500 mM Nacl, 0.2 µg/µl BSA, 20% glycerol, 1 mM DTT, pH 8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: UCHL1, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
Gene Symbol: UCHL1
Molecular Weight: 25.6 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: Ubiquitin carboxyl-terminal hydrolase isozyme L1
Protein Subtype: Proteases
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Neurobiology, Oncology
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols