Dimethyl Phthalate, 99%
Dimethyl Phthalate, 99%
Dimethyl Phthalate, 99%
Thermo Scientific Chemicals

Dimethyl Phthalate, 99%

CAS: 131-11-3 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 116642500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS131-11-3
IUPAC Name1,2-dimethyl benzene-1,2-dicarboxylate
Molecular FormulaC10H10O4
InChI KeyNIQCNGHVCWTJSM-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=98.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.5140 to 1.5160 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only