2,3-Dimethylquinoxaline, 97%
2,3-Dimethylquinoxaline, 97%
2,3-Dimethylquinoxaline, 97%
Thermo Scientific Chemicals

2,3-Dimethylquinoxaline, 97%

CAS: 2379-55-7 | C10H10N2 | 158.2 g/mol
Have Questions?
Catalog NumberQuantity
1167800505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116780050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2379-55-7
IUPAC Name2,3-dimethylquinoxaline
Molecular FormulaC10H10N2
InChI KeyFKHNZQFCDGOQGV-UHFFFAOYSA-N
SMILESCC1=C(C)N=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=96.0 %
Appearance (Color)Beige
Infrared spectrumConforms
Melting point102°C to 107°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only