Dimethyl sulfate, 99+%
Dimethyl sulfate, 99+%
Dimethyl sulfate, 99+%
Thermo Scientific Chemicals

Dimethyl sulfate, 99+%

CAS: 77-78-1 | C2H6O4S | 126.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
5 L
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116821000
Price (EUR)
28,65
Online exclusive
31,70
Save 3,05
Each
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
28,65
Online exclusive
31,70
Save 3,05
Each
Chemical Identifiers
CAS77-78-1
IUPAC Namedimethyl sulfate
Molecular FormulaC2H6O4S
InChI KeyVAYGXNSJCAHWJZ-UHFFFAOYSA-N
SMILESCOS(=O)(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to light brown
Appearance (Form)Oily liquid
Infrared spectrumConforms
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Titration99.0 to 101.0 % (Hydrolysis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only