2,4-Dinitroaniline, 99%
2,4-Dinitroaniline, 99%
2,4-Dinitroaniline, 99%
Thermo Scientific Chemicals

2,4-Dinitroaniline, 99%

CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1169300505 gGlass bottle
Catalog number 116930050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS97-02-9
IUPAC Name2,4-dinitroaniline
Molecular FormulaC6H5N3O4
InChI KeyLXQOQPGNCGEELI-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water8 to 15 % (K.F.)
Appearance (Form)Crystalline powder
Melting point176°C to 180°C
Appearance (Color)Yellow to yellow-green to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only