Glasgow's MEM (GMEM)
Gibco™

Glasgow's MEM (GMEM)

Glasgow's MEM (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's Minimal Essential Medium.Read more
Have Questions?
Catalog NumberQuantity
11710035500 mL
Catalog number 11710035
Price (USD)
52.65
Online exclusive
57.00
Save 4.35
Each
Quantity:
500 mL
Customize this product
Price (USD)
52.65
Online exclusive
57.00
Save 4.35
Each

Glasgow's MEM (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's Minimal Essential Medium. It was used to study the genetic factors that affected cell competence. Glasgow's MEM was developed for use with kidney cell lines, such as BHK-21.

This Glasgow's MEM is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red• Tryptose phosphate broth


The complete formulation is available.

Using Glasgow's MEM
Glasgow's MEM is unique from other media as it contains twice the concentration of amino acids and vitamins compared to the original Basal Medium Eagle, and is used without serum. Glasgow's MEM was originally formulated with 10% tryptose phosphate broth. Glasgow's MEM contains no proteins, lipids, or growth factors. Therefore, Glasgow's MEM requires supplementation with 10% tryptose phosphate broth. Glasgow's MEM uses a sodium bicarbonate buffer system (2.75 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineBHK-21
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMEM (Minimum Essential Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Phenol Red
Without AdditivesNo HEPES, No Sodium Pyruvate, No Tryptose Phosphate Broth
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.