Diphenylacetic Acid, 99+%
Diphenylacetic Acid, 99+%
Diphenylacetic Acid, 99+%
Thermo Scientific Chemicals

Diphenylacetic Acid, 99+%

CAS: 117-34-0 | C14H12O2 | 212.25 g/mol
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 117155000
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS117-34-0
IUPAC Name2,2-diphenylacetic acid
Molecular FormulaC14H12O2
InChI KeyPYHXGXCGESYPCW-UHFFFAOYSA-N
SMILESOC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Melting point146°C to 149°C
Appearance (Color)White to off-white to cream
Titration with NaOH>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only