Diphenyl carbonate, 99%
Diphenyl carbonate, 99%
Diphenyl carbonate, 99%
Thermo Scientific Chemicals

Diphenyl carbonate, 99%

CAS: 102-09-0 | C13H10O3 | 214.22 g/mol
数量:
100 g
1 kg
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 117230025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS102-09-0
IUPAC Namediphenyl carbonate
Molecular FormulaC13H10O3
InChI KeyROORDVPLFPIABK-UHFFFAOYSA-N
SMILESO=C(OC1=CC=CC=C1)OC1=CC=CC=C1
さらに表示
Appearance (Color)White
Infrared spectrumConforms
GC>=98.5 %
Melting point78°C to 81°C
Appearance (Form)Crystalline powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only