Diphenyl chlorophosphate, 98%
Diphenyl chlorophosphate, 98%
Diphenyl chlorophosphate, 98%
Thermo Scientific Chemicals

Diphenyl chlorophosphate, 98%

CAS: 2524-64-3 | C12H10ClO3P | 268.63 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
產品號碼 117245000
價格 (TWD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2524-64-3
IUPAC Namediphenyl phosphorochloridate
Molecular FormulaC12H10ClO3P
InChI KeyBHIIGRBMZRSDRI-UHFFFAOYSA-N
SMILESClP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5485 to 1.5505 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only