2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
Thermo Scientific Chemicals

2,2-Diphenylethylamine, 96%

CAS: 3963-62-0 | C14H15N | 197.28 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 117250050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3963-62-0
IUPAC Name2,2-diphenylethan-1-amine
Molecular FormulaC14H15N
InChI KeyRXMTUVIKZRXSSM-UHFFFAOYSA-N
SMILESNCC(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
GC>=95 %
Appearance (Form)Solid
Melting point43°C to 49°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only