CellsDirect Resuspension &amp; Lysis Buffers<i> includes 10 ml Resuspension Buffer &amp; 1 ml Lysis Buffer</i>
Invitrogen™

CellsDirect Resuspension & Lysis Buffers includes 10 ml Resuspension Buffer & 1 ml Lysis Buffer

These 2 buffers are to be used only as a complement to the SuperScript® III Platinum® CellsDirect Kits when largeRead more
Have Questions?
Catalog NumberQuantity
117390101 kit
Catalog number 11739010
Price (USD)
1,120.00
Each
Quantity:
1 kit
Price (USD)
1,120.00
Each
These 2 buffers are to be used only as a complement to the SuperScript® III Platinum® CellsDirect Kits when large number of samples are processed. See manual of SuperScript® III Platinum® CellsDirect Kits for protocol recommendations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineCellsDirect
Product TypeCell Lysis Buffer
Quantity1 kit
Shipping ConditionDry Ice
Concentration2X
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).