Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P
Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P
Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P
Thermo Scientific Chemicals

Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P

CAS: 4712-55-4 | C12H11O3P | 234.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 117395000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4712-55-4
IUPAC Nameoxodiphenoxy-λ⁵-phosphanylium
Molecular FormulaC12H10O3P
InChI KeyCDXVUROVRIFQMV-UHFFFAOYSA-N
SMILESO=[P+](OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Acid value=<15 mg KOH/g
Color scale=<50 APHA
Refractive index1.5565 to 1.5585 (20°C, 589 nm)
Specific gravity(25/15.5°C) 1.2 to 1.22
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only