Interested in a bulk or custom version of this product? Request pricing.

PDE1B Recombinant Human Protein, N-His.GST Tag

Catalog number: 11746H20B25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Phosphodiesterase 1B, Calmodulin-Dependent (PDE1B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human PDE1B long isoform (Q01064-1) (Met 1-Asp 536) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in 50 mM Tris, 100 mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Lyophilized
Gene Aliases: PDE1B, PDE1B1, PDES1B
Gene ID: 5153
Gene Symbol: PDE1B
Molecular Weight: 89.2 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Full Length, Recombinant
Protein Name: phosphodiesterase 1B, calmodulin-dependent
Protein Tag or Fusion: GST Tag, His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols

Recommended products