2,2'-Dipyridyl, 99+%
2,2'-Dipyridyl, 99+%
2,2'-Dipyridyl, 99+%
Thermo Scientific Chemicals

2,2'-Dipyridyl, 99+%

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 117500100
Price (USD)
54.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.00
Each
Chemical Identifiers
CAS366-18-7
IUPAC Name2,2'-bipyridine
Molecular FormulaC10H8N2
InChI KeyROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point69°C to 72°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only