n-Dodecane, 99%, pure
n-Dodecane, 99%, pure
n-Dodecane, 99%, pure
n-Dodecane, 99%, pure
Thermo Scientific Chemicals

n-Dodecane, 99%, pure

CAS: 112-40-3 | C12H26 | 170.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
250 mL
500 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 117590250
Price (TWD)
-
Request A Quote
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-40-3
IUPAC Namedodecane
Molecular FormulaC12H26
InChI KeySNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4210 to 1.4230 (20°C, 589 nm)
Color scale=<10 APHA
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only