SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument, 500 reactions - FAQs
SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument, 500 reactions - FAQs
View additional product information for SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument - FAQs (11760500, 11760100)
1 product FAQs found
Is there a SYBR Green Fast Master Mix currently available?
Yes. The following reagents can be used for fast cycling: Fast SYBR Green Master Mix, EXPRESS SYBR GreenER qPCR Supermix Universal, and SYBR GreenER qPCR SuperMix for ABI PRISM instruments.