ezDNase™ Enzyme
Invitrogen™
ezDNase™ Enzyme
Invitrogen™ ezDNase™ Enzyme is a recombinant double-strand-specific DNase for the fast removal of contaminating genomic DNA from RNA preparations. It자세히 알아보기
Have Questions?
카탈로그 번호수량
1176605150 reactions
카탈로그 번호 11766051
제품 가격(KRW)
325,000
온라인 행사
Ends: 30-Sep-2026
361,000
할인액 36,000 (10%)
Each
수량:
50 reactions
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
325,000
온라인 행사
Ends: 30-Sep-2026
361,000
할인액 36,000 (10%)
Each
Invitrogen™ ezDNase™ Enzyme is a recombinant double-strand-specific DNase for the fast removal of contaminating genomic DNA from RNA preparations. It cleaves phosphodiester bonds in double-stranded DNA to yield 2–8 bp oligonucleotides with 5’-phosphate and 3’-hydroxyl termini.
Features of ezDNase Enzyme include:
• Efficient and fast genomic DNA removal
• Highly specific—no impact on RNA, cDNA, or primers in RT reactions
ezDNase Enzyme's high specificity for double-stranded DNA enables efficient and fast genomic DNA removal without reduction in the quality or quantity of RNA or single-stranded DNA present in the reaction such as cDNA and primers. ezDNase Enzyme is heat-labile and so can be easily deactivated by heat treatment at moderate temperature (55°C). These features make ezDNase a superior choice for genomic DNA removal prior to reverse transcription reactions.
When used in combination with the SuperScript™ IV VILO™ Master Mix or other Invitrogen reverse transcription reagents, ezDNase Enzyme helps significantly reduce the possibility of cDNA synthesis being compromised by genomic DNA and reduce variation in RT-qPCR quantitation caused by RNA loss or damage during conventional DNase treatment.
For Research Use Only. Not for use in diagnostic procedures.
사양
함께 사용가능한 버퍼Reverse Transcription Reagents
용도(애플리케이션)RT-qPCR
제품 유형Enzyme
수량50 reactions
샘플 종류RNA
배송 조건On dry ice
농도10X
효소DNase
형태Liquid
제품라인ezDNase
Unit SizeEach
구성 및 보관
• ezDNase Enzyme, 50 μL
• ezDNase Buffer (10X), 100 μL
• Nuclease-free water, 1.25 mL
Store at -5 to -30°C.
자주 묻는 질문(FAQ)
How can I quantify the ssDNA in samples that contain both ssDNA and dsDNA, using the Qubit ssDNA Assay Kit (Cat. No. Q10212)?
Unfortunately, it is not possible to efficiently measure the ssDNA content in a sample that also contains dsDNA with the Qubit ssDNA Assay Kit (Cat. No. Q10212). The dye in the Qubit ssDNA Assay kit binds to both ssDNA and dsDNA.
It is possible to treat the sample with a DNase that only degrades dsDNA, such as theezDNase Enzyme (Cat. No. 11766051), and measure the ssDNA concentration afterward. However, if all dsDNA is not successfully digested, there is a risk of overestimating the ssDNA content.
The following article describes a PCR-based method to detect ssDNA in dsDNA samples:
Quantitative amplification of single-stranded DNA (QAOS) demonstrates that cdc13-1 mutants generate ssDNA in a telomere to centromere direction
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Does the TaqMan Cells-to-CT Express Kit contain reagents for the removal of genomic DNA (gDNA)?
Yes, the TaqMan Cells-to-CT Express Kit includes Express ezDNase which can be added to the Express Lysis Solution for removal of gDNA during cell lysis. Express ezDNase is a double-strand specific, heat-labile DNase that will only digest gDNA, making it compatible with the downstream reverse transcription reaction. The enzyme is automatically inactivated during a heat kill step included in the reverse transcription program. To avoid the detection of gDNA, we recommend using a TaqMan Gene Expression Assay specifically designed to span an intron-exon boundary. Such assays are designated with an "_m1" suffix. More information can be found here.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
In your SuperScript IV RT protocols, there is no ezDNase inactivation step. Will ezDNase be active to affect RNA or the RT reaction?
The Invitrogen ezDNase Enzyme is a novel DNase that is highly specific for double-stranded DNA. It has no activity on single-stranded DNA in RT reactions (primers or probes), or on RNA. The enzyme is also thermolabile—it is inactivated quickly at temperatures typical for the SuperScript IV RT reaction (e.g., 50 degrees C). The additional inactivation step is therefore not required for most RT-qPCR applications. If the RNA sample is to be used for RT-PCR of fragments ≥3 kb, incubate the sample for 5 minutes at 55 degrees C in the presence of 10 mM DTT to inactivate the enzyme.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Which SuperScript IV RT format do you recommend for real-time PCR applications?
For RT-qPCR applications we recommend using the Invitrogen SuperScript IV VILO Master Mix. The cDNA synthesis reaction setup with this master mix requires fewer pipetting steps and therefore reduces variation in the data. SuperScript IV RT, as a component of the master mix, offers the highest efficiency of cDNA synthesis step compared to competitors’ products.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Are there any significant changes in the SuperScript IV RT protocol compared to the SuperScript III RT protocol?
The only difference is that the incubation time for the reverse transcription reaction has been reduced to 10 minutes and inactivation time has been reduced to 5 minutes at 85 degrees C.