Dotriacontane, 97%
Dotriacontane, 97%
Dotriacontane, 97%
Thermo Scientific Chemicals

Dotriacontane, 97%

CAS: 544-85-4 | C32H66 | 450.88 g/mol
数量:
5 g
25 g
100 g
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 117690250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS544-85-4
IUPAC Namedotriacontane
Molecular FormulaC32H66
InChI KeyQHMGJGNTMQDRQA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
さらに表示
Appearance (Form)Glistening flakes
Melting point67°C to 72°C
Appearance (Color)White to light grey
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only