Emodin, 95%, tech.
Emodin, 95%, tech.
Emodin, 95%, tech.
Thermo Scientific Chemicals

Emodin, 95%, tech.

CAS: 518-82-1 | C15H10O5 | 270.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 117755000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS518-82-1
IUPAC Name1,3,8-trihydroxy-6-methyl-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC15H10O5
InChI KeyRHMXXJGYXNZAPX-UHFFFAOYSA-N
SMILESCC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=94 %
Infrared spectrumConforms
Appearance (Color)Orange-red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only