Ergosterol, 98%
Ergosterol, 98%
Ergosterol, 98%
Thermo Scientific Chemicals

Ergosterol, 98%

CAS: 57-87-4 | C28H44O | 396.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 117810050
Price (EUR)
54,65
Online exclusive
60,30
Save 5,65
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
54,65
Online exclusive
60,30
Save 5,65
Each
Chemical Identifiers
CAS57-87-4
IUPAC Name(1R,3aR,7S,9aR,9bS,11aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC28H44O
InChI KeyDNVPQKQSNYMLRS-APGDWVJJSA-N
SMILESCC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Crystalline powder or needles
Specific optical rotation-105° to -135° (20°C, 589 nm) (c=1, CHCl3)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only