1,2-Ethanedithiol, 98+%
1,2-Ethanedithiol, 98+%
1,2-Ethanedithiol, 98+%
Thermo Scientific Chemicals

1,2-Ethanedithiol, 98+%

CAS: 540-63-6 | C2H6S2 | 94.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 117850010
Price (TWD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS540-63-6
IUPAC Nameethane-1,2-dithiol
Molecular FormulaC2H6S2
InChI KeyVYMPLPIFKRHAAC-UHFFFAOYSA-N
SMILESSCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to slightly colored
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only