2-Ethoxybenzamide, 97%
2-Ethoxybenzamide, 97%
2-Ethoxybenzamide, 97%
Thermo Scientific Chemicals

2-Ethoxybenzamide, 97%

CAS: 938-73-8 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number 117870050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS938-73-8
IUPAC Name2-ethoxybenzamide
Molecular FormulaC9H11NO2
InChI KeySBNKFTQSBPKMBZ-UHFFFAOYSA-N
SMILESCCOC1=CC=CC=C1C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point129°C to 134°C
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only