2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
Thermo Scientific Chemicals

2(2-Ethoxyethoxy)ethanol, 98+%

CAS: 111-90-0 | C6H14O3 | 134.17 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
檢視全部
Glass Bottle
產品號碼 117890010
價格 (TWD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
化學識別
CAS111-90-0
IUPAC Name2-(2-ethoxyethoxy)ethan-1-ol
Molecular FormulaC6H14O3
InChI KeyXXJWXESWEXIICW-UHFFFAOYSA-N
SMILESCCOCCOCCO
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4250 to 1.4280 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only