2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
Thermo Scientific Chemicals

2(2-Ethoxyethoxy)ethanol, 98+%

CAS: 111-90-0 | C6H14O3 | 134.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 117890025
Price (USD)
157.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
157.00
Each
Chemical Identifiers
CAS111-90-0
IUPAC Name2-(2-ethoxyethoxy)ethan-1-ol
Molecular FormulaC6H14O3
InChI KeyXXJWXESWEXIICW-UHFFFAOYSA-N
SMILESCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4250 to 1.4280 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only