2-Ethoxyethyl Ether, 98+%, Extra Pure
2-Ethoxyethyl Ether, 98+%, Extra Pure
2-Ethoxyethyl Ether, 98+%, Extra Pure
Thermo Scientific Chemicals

2-Ethoxyethyl Ether, 98+%, Extra Pure

CAS: 112-36-7 | C8H18O3 | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 117900010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-36-7
IUPAC Name1-ethoxy-2-(2-ethoxyethoxy)ethane
Molecular FormulaC8H18O3
InChI KeyRRQYJINTUHWNHW-UHFFFAOYSA-N
SMILESCCOCCOCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acidity (CH3COOH)=<0.015 %
Appearance (Form)Clear liquid
GC>=98.0 %
Residue after evaporation=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only