4-Ethylaniline, 99+%
4-Ethylaniline, 99+%
4-Ethylaniline, 99+%
Acros Organics

4-Ethylaniline, 99+%

CAS: 589-16-2 | C8H11N | 121.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
118065000
also known as 11806
500 gGlass bottle
Catalog number 118065000
also known as 11806
HSN 29214290
Price (INR)
67,558.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
67,558.00
Each
Chemical Identifiers
CAS589-16-2
IUPAC Name4-ethylaniline
Molecular FormulaC8H11N
InChI KeyHRXZRAXKKNUKRF-UHFFFAOYSA-N
SMILESCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to red-brown
Infrared spectrumConforms
Refractive index1.5532 to 1.5552 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only