Ethyl Benzoate, 99+%
Ethyl Benzoate, 99+%
Ethyl Benzoate, 99+%
Thermo Scientific Chemicals

Ethyl Benzoate, 99+%

CAS: 93-89-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 118090025
Price (USD)
108.65
Online exclusive
121.00
Save 12.35
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
108.65
Online exclusive
121.00
Save 12.35
Each
Chemical Identifiers
CAS93-89-0
IUPAC Nameethyl benzoate
Molecular FormulaC9H10O2
InChI KeyMTZQAGJQAFMTAQ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=99.0 %
Specific gravity(25/25°C) 1.043 to 1.047
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only