You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Baths
▸
Bath Accessories
▸
Concentric Ring Covers for Lindberg/Blue M™ Multi-Purpose Circulating Baths
Actual product may vary
Thermo Scientific™
Concentric Ring Covers for Lindberg/Blue M™ Multi-Purpose Circulating Baths
Use these 2–, 6– or 8–ring Concentric-Ring Covers with Lindberg Blue/M baths.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
118091
WB1110
118092
Models WB1120 and RWB3220
118093
WB1130
118094
WB1140
4 Options
Catalog number
118091
Price (MXN)
-
For Use With (Equipment):
WB1110
Use Thermo Scientific™ 2–, 6– or 8–ring Concentric-Ring Covers with Lindberg Blue/M baths.
Warranty varies by country, please contact us for details
Specifications
Description
2 rings
For Use With (Equipment)
WB1110
Unit Size
Each