Upgrade Kits
Product Image
Thermo Scientific™
Upgrade Kits
Take advantage of technology improvements with these in-field Upgrade Kits for the Thermo Scientific™ ELEMENT™ HR-ICP-MS analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
1181030Retrofit-Kit for Plug-in Slit
1156420Peltier Cooling ICP
1179730SEV ICP2-V2 PLUG IN Upgrade
1180080SEM ICP2-V2 Upgrade Kit
1289760Pump for Jet Interface
Catalog number 1181030
Price (BRL)
-
Includes:
Retrofit-Kit for Plug-in Slit
Enhance your Thermo Scientific™ ELEMENT™ HR-ICP-MS analyzer with these in-field retrofit Thermo Scientific™ Upgrade Kits to enable your HR-ICP-MS analysis capabilities to improve and progress in line with advances in technology.
Upgrade Kits for Improved Performance
  • Multiplier SEM Type II Detector; two specific retrofit kit versions, for either pre- or post-February 2003 instrument shipments
  • Peltier cooling for spray chamber; in-field retrofit kit
  • Plug-in slit kit; retrofit kit for ELEMENT 2 HR-ICP-MS model for the user-replaceable slit tongues
  • High capacity Interface Pump for Jet Interface; in-field upgrade
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS system
IncludesRetrofit-Kit for Plug-in Slit
TypeUpgrade Kit
Unit SizeEach