DL-Etil 2-bromopropionato, 99 %, Thermo Scientific Chemicals
DL-Etil 2-bromopropionato, 99 %, Thermo Scientific Chemicals
DL-Etil 2-bromopropionato, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Etil 2-bromopropionato, 99 %, Thermo Scientific Chemicals

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Número de catálogoCantidadEnvase
118141000100 gFrasco de vidrio
Número de catálogo 118141000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS535-11-5
IUPAC Nameethyl 2-bromopropanoate
Molecular FormulaC5H9BrO2
InChI KeyARFLASKVLJTEJD-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only