Ethyl butyrate, 99%
Ethyl butyrate, 99%
Ethyl butyrate, 99%
Thermo Scientific Chemicals

Ethyl butyrate, 99%

CAS: 105-54-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 118180010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS105-54-4
IUPAC Nameethyl butanoate
Molecular FormulaC6H12O2
InChI KeyOBNCKNCVKJNDBV-UHFFFAOYSA-N
SMILESCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3900 to 1.3940 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only