Ethyl carbazate, 97%
Ethyl carbazate, 97%
Ethyl carbazate, 97%
Thermo Scientific Chemicals

Ethyl carbazate, 97%

CAS: 4114-31-2 | C3H8N2O2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 118205000
Price (USD)
349.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
349.00
Each
Chemical Identifiers
CAS4114-31-2
IUPAC Nameethoxycarbohydrazide
Molecular FormulaC3H8N2O2
InChI KeyVYSYZMNJHYOXGN-UHFFFAOYSA-N
SMILESCCOC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)May have a pink hue
Infrared spectrumConforms
Melting point42°C to 48°C
Appearance (Color)White to almost white
Appearance (Form)Crystals and/or chunks or fused solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only