Sodium Pyruvate is commonly added to cell culture medium as an energy source in addition to glucose. Since the cells make sodium pyruvate as an intermediate metabolite in the glycolysis pathway, it is not necessary to supplement all cultures. However, if the cells have been grown in medium supplemented with sodium pyruvate, we recommend the supplementation is continued as the cells become accustomed to this addition and their growth will lag without it. Gibco® Sodium Pyruvate, powder, should be made into a 100 mM stock solution in water and filter sterilized. The final concentration of sodium pyruvate used in most cell culture medium is 1 mM.
Product Use For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 each
Inhalt und Lagerung
Storage conditions: 15 – 30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture