Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Thermo Scientific Chemicals

Ethylene carbonate, 99+%

CAS: 96-49-1 | C3H4O3 | 88.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 118410051
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS96-49-1
IUPAC Name1,3-dioxolan-2-one
Molecular FormulaC3H4O3
InChI KeyKMTRUDSVKNLOMY-UHFFFAOYSA-N
SMILESO=C1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point34°C to 37°C
Appearance (Form)Low melting crystalline solid
GC>=99.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
RUO – Research Use Only