Ethylenediamine, 99%
Ethylenediamine, 99%
Ethylenediamine, 99%
Thermo Scientific Chemicals

Ethylenediamine, 99%

CAS: 107-15-3 | C2H8N2 | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 118420010
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
71.90
Save 10.25
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
71.90
Save 10.25
Each
Chemical Identifiers
CAS107-15-3
IUPAC Nameethane-1,2-diamine
Molecular FormulaC2H8N2
InChI KeyPIICEJLVQHRZGT-UHFFFAOYSA-N
SMILESNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 % (Coulometric)
Appearance (Form)Liquid
Refractive index1.4555 to 1.4575 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only