Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure
Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure
Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure

Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, C10H12N2Na4O8.xH2O, CAS Number-194491-31-1 | CAS: 194491-31-1 | C10H12N2Na4O8 | 380.17 g/mol
Catalog NumberQuantity
1184400252.5 kg
Catalog number 118440025
Price (JPY)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS194491-31-1
IUPAC Nametetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12N2Na4O8
InChI KeyUEUXEKPTXMALOB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Complexometric97.5 to 102.5 % (on dry substance)
Loss on drying5 % to 15 % (250°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only